Alfa Aesar™ Titanium nitride sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 41112KS

Quantity 1 each

 View more versions of this product

Catalog No. AA41112KS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25583-20-4
    Formula Weight 61.91
    MDL Number MFCD00049596
    Chemical Name or Material Titanium nitride sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick

    Specifications

    Physical Form Hot Pressed Material
    Formula Weight 61.91
    MDL Number MFCD00049596
    CAS 25583-20-4
    Density 5.22g/mL
    RTECS Number XR2230000
    TSCA TSCA
    Color Gray
    Melting Point 2950°C
    Quantity 1 each
    Chemical Name or Material Titanium nitride sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
    Assay 99.5%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit