Alfa Aesar™ Titanium oxide, 99.9% (metals basis)

For vacuum deposition

Manufacturer: Alfa Aesar™ 4163518

Quantity 50g

 View more versions of this product

Catalog No. AA4163518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12065-65-5
    Linear Formula Ti3O5
    MDL Number MFCD00210650
    Synonym Trititanium pentoxide
    Chemical Name or Material Titanium oxide, Pellets, ≈8 to 9mm dia. x 7 to 9mm thick

    Specifications

    Physical Form Pellet
    CAS 12065-65-5
    Color Black
    Linear Formula Ti3O5
    MDL Number MFCD00210650
    Synonym Trititanium pentoxide
    Quantity 50g
    Melting Point 1800°C
    Density 4.20g/mL
    Chemical Name or Material Titanium oxide, Pellets, ≈8 to 9mm dia. x 7 to 9mm thick
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit