Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Titanium oxide, 99.9% (metals basis)

For vacuum deposition

Manufacturer:  Alfa Aesar™ 4163509

Quantity 10g

 View more versions of this product

Catalog No. AA4163509


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12065-65-5
MFCD00210650
Titanium oxide, Pellets, ≈8 to 9mm dia. x 7 to 9mm thick
Ti3O5
Trititanium pentoxide

Specifications

12065-65-5
Ti3O5
Trititanium pentoxide
Black
10g
99.9%, (metals basis)
Pellet
MFCD00210650
4.20g/mL
1800°C
Titanium oxide, Pellets, ≈8 to 9mm dia. x 7 to 9mm thick
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit