Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Titanium, plasma standard solution, Specpure™, Ti 10,000μg/mL

Click to view available options
Quantity:
50mL
100mL
500mL
MFCD00011264
MFCD00011264
UN Number UN3264
Concentration Ti in 5% HNO3/tr. HF
Physical Form Liquid
Grade Specpure™
Quantity 100mL
Chemical Name or Material Titanium plasma standard solution, Ti 10000μg/mL

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures