Alfa Aesar™ Titanium powder, -100 mesh, 99.4%

Manufacturer: Alfa Aesar™ 1038388

Quantity 2.5g

 View more versions of this product

Catalog No. AA1038388

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7440-32-6
    MDL Number MFCD00011264
    UN Number UN2546
    Chemical Name or Material Titanium powder

    Specifications

    Health Hazard 3 GHS P Statement
    P280-P235+P410-P420-P407-P413a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep cool.
    Protect from sunlight.
    Store away from other materials.
    Maintain air gap between stacks/pallets.
    Store bulk masses greater than 1 lb. at temperatures not exceeding 100°F.
    MDL Number MFCD00011264
    UN Number UN2546
    Odor Odorless
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H252
    Self-heating in large quantities
    may catch fire.
    DOT Information Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: TITANIUM POWDER, DRY
    TSCA TSCA
    Physical Form Powder
    CAS 7440-32-6
    Mesh Size −100 Mesh
    Color Gray
    RTECS Number XR1700000
    Quantity 2.5g
    Chemical Name or Material Titanium powder
    Assay 99.4%

    GHS H Statement
    H252
    Self-heating in large quantities
    may catch fire.

    GHS P Statement
    P280-P235+P410-P420-P407-P413a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep cool.
    Protect from sunlight.
    Store away from other materials.
    Maintain air gap between stacks/pallets.
    Store bulk masses greater than 1 lb. at temperatures not exceeding 100°F.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit