Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium Powder, -325 mesh, 99.5% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 42624D3

Quantity 2 x 1kg

 View more versions of this product

Catalog No. AA42624D3


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-32-6
99.5% (Metals basis)
Titanium
Powder
MFCD00011264

Specifications

2 x 1kg
Silver Gray powder
99.5% (Metals basis)
Odorless
Titanium
7440-32-6
Powder
MFCD00011264
Silver-gray

Safety and Handling

SDS & Safety Documents

Take precautionary measures against static discharges.

H251; Self-heating: may catch fire.

DOTInformation : DOT Class: 4.2, Packaging Group: II

EINECSNumber : 231-142-3

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit