Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Titanium Powder, dehydrided, -325 mesh, 99.99% (Metals basis), Alfa Aesar™
Click to view available options
Quantity:
25g
100g
Specifications
Quantity | 100g |
Appearance | Dark Gray powder |
Assay Percent Range | 99.99% (Metals basis) |
Mesh Size | -325 Mesh |
Odor | Odorless |
Physical Form | Dehydrided Powder |
Color | Gray |
Chemical Name or Material | Titanium |
Safety and Handling
P210-P222-P280-P335+P334-P370+P378q
H250-H500
DOTInformation : DOT Class: 4.2, Packaging Group: II
EINECSNumber : 231-142-3
RTECSNumber : XR1700000
TSCA : Yes
Recommended Storage : Ambient temperatures