Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Titanium powder, hydrided, -325 mesh, 99.99% (metals basis)
Click to view available options
Quantity:
25g
100g
Specifications
Quantity | 100g |
UN Number | UN2546 |
Mesh Size | −325 Mesh |
Odor | Odorless |
Physical Form | Powder |
Percent Purity | 99.99% |
Chemical Name or Material | Titanium powder, Hydrided |
Assay | (metals basis) |
Safety and Handling
GHS H Statement
H251
Self-heating: may catch fire.
P210-P222-P280-P335+P334-P370+P378q
H250-H500
DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: TITANIUM POWDER, DRY
EINECSNumber : 231-726-8
RTECSNumber : XR1700000
TSCA : Yes
Recommended Storage : Ambient temperatures