Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

Click to view available options
Quantity:
100mL
500mL
Packaging:
Poly Bottle
7440-32-6,7722-84-1,7732-18-5,7722-84-1
Ti
47.867
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
CHEBI:33341
titanium
[Ti]
7440-32-6,7722-84-1,7732-18-5,7722-84-1
Ti
47.867
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
CHEBI:33341
titanium
[Ti]
Concentration 990 to 1010ppm
Physical Form Liquid
Grade Certified
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED TITANIUM STANDARD
Vapor Pressure 14mmHg at 20°C
Density 0.7g/cm³
Melting Point 0°C
Boiling Point 100°C
Color Red
Quantity 500mL
Chemical Name or Material Titanium, Reference Standard Solution, 1000ppm ±1%
Show More Show Less

DOTInformation : DOT Class 8, : Corrosive