Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.7% (metals basis)

Click to view available options
Quantity:
1 ea.
7440-32-6
Ti
47.867
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]
7440-32-6
Ti
47.867
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet,alloy,cp,vt1,c.p.,contimet 30,jistp28,titanium-125,ii hydride,titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]
Quantity 1 ea.
Odor Odorless
Percent Purity 99.7%
Chemical Name or Material Titanium sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay (metals basis)

EINECSNumber : 231-142-3

TSCA : Yes

Recommended Storage : Ambient temperatures