Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium Standard, 1mL = 1mg Ti, 1000ppm Ti, Ricca Chemical
SDP

Catalog Number p-3252588
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Glass amber
7440-32-6, 7647-01-0, 7732-18-5
Ti
47.87
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
CHEBI:33341
titanium
[Ti]
7440-32-6, 7647-01-0, 7732-18-5
Ti
47.87
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
CHEBI:33341
titanium
[Ti]
Physical Form Liquid
Packaging Glass amber
Equivalence 1 mL = 1 mg Ti
Color Colorless to Purple
Quantity 120 mL
Concentration or Composition Notes 1,000ppm Ti
Chemical Name or Material Titanium Standard
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit