Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Titanium Std., 10,000μg/mL in water w/Dilute Nitric and Trace HF Acids, Ultra Scientific
Click to view available options
Quantity:
125 mL
Specifications
Quantity | 125 mL |
Concentration or Composition (by Analyte or Components) | 10, 000μg/mL |
Physical Form | Liquid |
Chemical Name or Material | Titanium Standard, 10, 000μg/mL |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction