Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium Tetrachloride RS, ACS Reagent Solution, Ricca Chemical

Click to view available options
Quantity:
500mL
Packaging:
Glass amber
7647-01-0,7732-18-5,7550-45-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
7647-01-0,7732-18-5,7550-45-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof
313
CHEBI:17883
chlorane
Cl
CAS Min % 49.0,47.14,1.53
CAS Max % 51.0,49.07,1.59
Physical Form Liquid
Grade ACS Reagent
Packaging Glass amber
Color Colorless to Yellow
Quantity 500mL
Chemical Name or Material Titanium Tetrachloride RS

ShelfLife : 12 months