Alfa Aesar™ Titanium Thinfoil, 0.0127mm (0.0005 in.) thick, 99.6% (metals basis)

Manufacturer: Alfa Aesar™ 442436Z

Quantity 90x500mm

 View more versions of this product

Catalog No. AA442436Z

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-32-6
    MDL Number MFCD00011264
    Chemical Name or Material Titanium Thinfoil, 0.0127mm (0.0005 in.) thick, Light Tight
    Grade Commercial Pure Grade 4

    Specifications

    Physical Form Thin Foil
    TSCA TSCA
    MDL Number MFCD00011264
    CAS 7440-32-6
    Odor Odorless
    Weight ≈0.515g/90x100mm
    Quantity 90x500mm
    Chemical Name or Material Titanium Thinfoil, 0.0127mm (0.0005 in.) thick, Light Tight
    Assay 99.6%, (metals basis)
    Grade Commercial Pure Grade 4

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit