Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanocene Bis(trifluoromethanesulfonate) 97.0+%, TCI America™

Click to view available options
Quantity:
100mg
1g
76262-87-8
C12H10F6O6S2Ti
MFCD00192505
Bis(cyclopentadienyl)titanium(IV) Bis(trifluoromethanesulfonate)
76262-87-8
C12H10F6O6S2Ti
MFCD00192505
Bis(cyclopentadienyl)titanium(IV) Bis(trifluoromethanesulfonate)
Color Undesignated
Quantity 1g
Formula Weight 476.18
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (T)
Chemical Name or Material Titanocene Bis(trifluoromethanesulfonate)