Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Titer Plate Shakers

Thermo Scientific™ Titer Plate Shakers have been discontinued.

Supplier:  Thermo Scientific™ 4625Q

 View more versions of this product

Catalog No. 14-271-9

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Thermo Scientific™ Titer Plate Shakers have been discontinued. Please consider the Thermo Scientific™ Compact Digital Microplate Shaker as its replacement.

Features:
  • Solid-state, variable speed control from 40 to 1100rpm with a 0.12″ (0.3cm) circular orbit
  • Electronic timer can be set for continuous operation or timed shaking up to 5 min. (adjustable in 30-second increments)
  • Independent start switch activates shaker for preset speed and time for repeatability in tests
  • Automatic shutoff
  • Platform has 0.38″ (0.9cm) sides and holds from one to four 96-well standard or deep-well plates
  • Two retaining springs and nonslip rubber mat hold plates securely
  • Four platform finger slots for easy plate removal
  • Rugged, enameled steel shaker body with four nonskid rubber feet
TRUSTED_SUSTAINABILITY
Specifications

Specifications

40 to 1100 rpm
120 V
1 A
32° to 104°F
Rubber, Steel
11 kg
Continuous or timed from 30 sec. to 5 min., adjustable in 30-sec. increments
120 V, 60 Hz, 1 A
0°C to 40°C
CE, UL Listed
60 Hz
25 lb.
20 to 80%, Non-condensing
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit