Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titriplex™ III (ethylenedinitrilotetraacetic Acid disodium salt dihydrate) EMPROVE™ ESSENTIAL Ph Eur, BP, ChP, JP, USP, MilliporeSigma™

Click to view available options
Quantity:
25 kg
CAS 6381-92-6
pH 4 to 5 (50 g/L, H2O, 20°C)
Melting Point 110°C
Molecular Formula C10H14N2Na2O8 * 2H2O
Packaging Plastic Drum
Quantity 25 kg
Synonym Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetraacetic acid disodium salt
Molecular Weight (g/mol) 372.24