Learn More
TJAP1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 177341AP
Catalog No. 50-173-1755
Add to cart
Description
Description
This gene encodes a tight junction-associated protein. Incorporation of the encoded protein into tight junctions occurs at a late stage of formation of the junctions. The encoded protein localizes to the Golgi and may function in vesicle trafficking. Alternatively spliced transcript variants have been described. A related pseudogene exists on the X chromosome.
Specifications
Specifications
TJAP1
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5JTD0
PILT, Tight junction protein 4, TJAP1, TJP4
Rabbit
IgG
150 μL
-20°C
Primary
93643
Western Blot
Unconjugated
TJAP1
Liquid
TJAP1
TJAP1 Fusion Protein Ag12174
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human