Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TK15 Epitope Tag Antibody (TK 15), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250335AF700
Description
TK15 Epitope Tag Monoclonal antibody specifically detects TK15 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
TK15 Epitope Tag | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Recombinant full length Xenopus Orc1p protein | |
0.1 mL | |
Primary | |
Purified | |
Epitope Tag |
Western Blot, Immunocytochemistry, Immunofluorescence | |
TK 15 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction