Learn More
Azzota Corp TKI-258 DFS Item
Manufacturer:  Azzota Corp C1317
TKI-258 (Dovitinib) is an orally bioavailable FGFR3 inhibitor, which strongly binds to FGFR3 and inhibits its phosphorylation, which may result in the inhibition of tumor cell proliferation and the induction of tumor cell death.
Catalog No. 50-161-3189