Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TKN Digestion Reagent with Copper Sulfate, Reagents

Catalog No. p-200021429
Click to view available options
Quantity:
4 L
10 L

TKN Digestion Reagent with Copper Sulfate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5, 7664-93-9, 7778-80-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Total Kjeldahl Nitrogen Digestion Reagent
IUPAC Name water
SMILES O

Specifications

Color Blue
Quantity 10 L
Boiling Point 100°C
Packaging Cubitainer
Physical Form Liquid
Grade Reagent
Chemical Name or Material TKN Digestion Reagent with Copper Sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.