Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TL1A/TNFSF15, Rabbit anti-Mouse, Clone: TAN2-2, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen TL1A/TNFSF15
Clone TAN2-2
Applications Flow Cytometry, Immunofluorescence, Blocking Assay
Host Species Rabbit
Target Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB016552
View Documents
Novus Biologicals
NBP2811200.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB016553
View Documents
Novus Biologicals
NBP2811200.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

TL1A/TNFSF15 Monoclonal antibody specifically detects TL1A/TNFSF15 in Mouse Samples. It is validated for Flow Cytometry, Immunofluorescence, Blocking Assay.
Specifications

Specifications

TL1A/TNFSF15
Flow Cytometry, Immunofluorescence, Blocking Assay
Mouse
Primary
TAN2-2
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit