Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLE1, Mouse anti-Human, Alexa Fluor 700, Clone: TLE1/2062, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279914AF700
Description
TLE1 Monoclonal antibody specifically detects TLE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
TLE1 | |
TLE1/2062 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
TLE1 | |
Recombinant human TLE1 fragment (aa 175-338) (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
E(Sp1) homolog, enhancer of split groucho 1, ESG, ESG1Enhancer of split groucho-like protein 1, GRG1, transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 1, homolog of Drosophila E(sp1), transducin-like enhancer protein 1 | |
Mouse | |
IgG2a Kappa | |
0.1 mL | |
Signal Transduction | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title