Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLE1 Mouse anti-Human, Clone: TLE1/2085, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | TLE1 |
---|---|
Clone | TLE1/2085 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122928
|
Novus Biologicals
NBP30772720UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
TLE1 Monoclonal specifically detects TLE1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
TLE1 | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Signal Transduction | |
10 mM PBS with 0.05% BSA | |
7088 | |
IgG2a κ | |
Protein A or G purified |
TLE1/2085 | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
E(Sp1) homolog, enhancer of split groucho 1, ESG, ESG1Enhancer of split groucho-like protein 1, GRG1, transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 1, homolog of Drosophila E(sp1), transducin-like enhancer protein 1 | |
Recombinant fragment (aa 175-338) of human TLE1 (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title