Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLE1, Mouse anti-Human, FITC, Clone: TLE1/2051, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279913F

Catalog No. NB012439


Add to cart

Description

Description

TLE1 Monoclonal antibody specifically detects TLE1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

TLE1
TLE1/2051
Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
TLE1
Recombinant human TLE1 fragment (aa 175-338) (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C in the dark.
Monoclonal
Immunohistochemistry, Immunohistochemistry (Paraffin)
FITC
PBS with 0.05% Sodium Azide
E(Sp1) homolog, enhancer of split groucho 1, ESG, ESG1Enhancer of split groucho-like protein 1, GRG1, transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 1, homolog of Drosophila E(sp1), transducin-like enhancer protein 1
Mouse
IgG2a Kappa
0.1 mL
Signal Transduction
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit