Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TLE2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528360
Description
Recommended positive controls: H1299. Predicted reactivity: Mouse (97%), Rat (96%), Zebrafish (89%), Xenopus laevis (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TLE2 (Transducin Like Enhancer Of Split 2) is a Protein Coding gene. Among its related pathways are Signaling by Wnt and Wnt / Hedgehog / Notch. Gene Ontology (GO) annotations related to this gene include transcription corepressor activity. An important paralog of this gene is TLE1.
Specifications
TLE2 | |
Polyclonal | |
Unconjugated | |
TLE2 | |
enhancer of split groucho-like protein 2; ESG; ESG2; ESG-2; GRG2; TLE2; transducin like enhancer of split 2; transducin-like enhancer of split 2 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 2, homolog of Drosophila E(sp1); transducin-like enhancer protein 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7089 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.75 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q04725 | |
TLE2 | |
Recombinant fragment corresponding to a region within amino acids 529 and 743 of Human TLE2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction