Learn More
TLE3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 220941AP
Catalog No. 50-173-3722
Add to cart
Description
Description
The TLE genes are differentially expressed and encode nuclear proteins, consistent with the presence of sequence motifs associated with nuclear functions. These genes are the human homologues of Drosophila groucho, a member of the Notch signaling pathway which controls a number of different cell-fate choices in invertebrates and vertebrates.
Specifications
Specifications
TLE3
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q04726, Q08122, Q9JIT3
ESG, ESG3, GRG3, HsT18976, KIAA1547, TLE3
Rabbit
IgG
150 μL
-20°C
Primary
21887, 7090, 84424
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
TLE3
Liquid
TLE3
TLE3 Fusion Protein Ag17300
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat