Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLK1 Rabbit anti-Human, Alexa Fluor 488, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP298826AF488

Catalog No. NB106043


Add to Cart

Description

Description

TLK1 Polyclonal specifically detects TLK1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

TLK1
Polyclonal
50 mM Sodium Borate
Rabbit
Antigen Affinity-purified
RUO
Primary
Human
Purified
Western Blot, Immunofluorescence
Alexa Fluor 488
EC 2.7.11, EC 2.7.11.1, KIAA0137serine/threonine-protein kinase tousled-like 1, PKU-beta, SNARE protein kinase SNAK, tousled-like kinase 1serine threonine protein kinase
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human TLK1.
0.1 mL
Apoptosis, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Repair, Tumor Suppressors
9874
Store at 4°C in the dark.
IgG
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit