Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLK2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | TLK2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TLK2 Polyclonal specifically detects TLK2 in Human samples. It is validated for Western Blot.Specifications
TLK2 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication | |
EC 2.7.11, EC 2.7.11.1, MGC44450, PKU-ALPHA, serine/threonine-protein kinase tousled-like 2, tousled-like kinase 2serine/threonine kinase | |
TLK2 | |
IgG | |
82 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q86UE8 | |
11011 | |
Synthetic peptides corresponding to TLK2(tousled-like kinase 2) The peptide sequence was selected from the N terminal of TLK2. Peptide sequence VSAQQNSPSSTGSGNTEHSCSSQKQISIQHRQTQSDLTIEKISALENSKN. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title