Learn More
TLK2 Mouse anti-Human, Clone: OTI2F9, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805461
Catalog No. 50-166-7439
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TLK2, a member of the Ser/Thr protein kinase family, is rapidly and transiently inhibited by phosphorylation following the generation of DNA double-stranded breaks during S-phase. This is cell cycle checkpoint and ATM-pathway dependent and appears to regulate processes involved in chromatin assembly. The TLK2 enzyme is cell-cycle regulated, with maximal activity in S-phase. It is inactivated by phosphorylation at Ser-750, potentially by CHK1. TLK2 heterodimerizes with TLK1. This nuclear protein is widely expressed, with presence in fetal placenta, liver, kidney, pancreas, heart and skeletal muscle tissues, and in several adult cell lines.
Specifications
Specifications
TLK2
Monoclonal
Unconjugated
TLK2
Lyophilized
TLK2
Human recombit protein fragment corresponding to amino acids 1-296 of human TLK2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI2F9
PBS with 8% trehalose and no preservative; pH 7.3
Q86UE8
HsHPK, PKU-ALPHA
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
11011