Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR1 Polyclonal Antibody
TLR1 Polyclonal Antibody
Supplier: Thermo Scientific PA580135
Description
TLR1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P), Flow

Specifications
TLR1 | |
Polyclonal | |
Unconjugated | |
TLR1 | |
CD281; KIAA0012; rsc786; TIL; TIL. LPRS5; TLR1; toll like receptor 1; Toll/interleukin-1 receptor-like protein; toll-like receptor 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21897, 305354, 7096 | |
-20°C | |
Lyophilized |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
Q15399, Q9EPQ1 | |
TLR1 | |
E. coli-derived human TLR1 recombinant protein (Position: F350-D404). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction