Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
TLR10 (CD290) Mouse anti-Human, BV421, Clone: 3C10C5, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566232
Catalog No. BDB566232
Description
Description
TLR10 (CD290) Monoclonal antibody specifically detects TLR10 (CD290) in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
TLR10 (CD290)
3C10C5
Brilliant Violet 421
Purified
Murine
Mouse BALB/c IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Toll-like receptor 10; CD290
Human TLR10 Recombinant Protein
25μg
Cell Biology
Primary
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.