Learn More
TLR10 Mouse anti-Human, Alexa Fluor™ 350, Clone: 28M7A3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227273AF350
Catalog No. NP227273A30
Add to cart
Description
Description
TLR10 Monoclonal antibody specifically detects TLR10 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TLR10
28M7A3
Flow Cytometry
CD290, CD290 antigen, MGC104967, MGC126398, MGC126399, toll-like receptor 10
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
TLR10
A portion of amino acids 300-350 of human TLR10 was used as immunogen for this antibody.
0.1 mL
Primary
81793