Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR10 Mouse anti-Human, Alexa Fluor™ 532, Clone: 28M7A3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP227273AF532

Catalog No. NP227273A32


Add to cart

Description

Description

TLR10 Monoclonal antibody specifically detects TLR10 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

TLR10
28M7A3
Flow Cytometry
Conjugated, Purified
TLR10
A portion of amino acids 300-350 of human TLR10 was used as immunogen for this antibody.
Protein G purified
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary
81793.0
Flow Cytometry
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CD290, CD290 antigen, MGC104967, MGC126398, MGC126399, toll-like receptor 10
Mouse
IgG2a Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit