Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR10 Rabbit anti-Chimpanzee, Human, Non-human Primate, Rat, Polyclonal, FabGennix

Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc TLR1001AP

Description
TLR10 Polyclonal antibody specifically detects TLR10 in Chimpanzee, Human, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
TLR10 | |
Polyclonal | |
Unconjugated | |
TLR10 | |
CD290; MGC104967; MGC126398; MGC126399; TLR10; toll like receptor 10; toll-like receptor 10; UNQ315/PRO358 | |
Synthetic peptide taken within amino acid region 380-430 on human TLR10 protein. | |
200 μL | |
Primary | |
Chimpanzee, Human, Non-human Primate, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q9BXR5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
305355, 461166, 81793 | |
-20°C | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title