Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR11 Rabbit anti-Chimpanzee, Human, Non-human Primate, Biotin, Polyclonal, FabGennix

Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc TLR11BIOTIN

Description
TLR11 Polyclonal antibody specifically detects TLR11 in Chimpanzee, Human, Non-human Primate samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
TLR11 | |
Polyclonal | |
Biotin | |
Gm1365; Gm287; RGD1564460; RP23-392K10.4; Tlr11; Tlr12; Toll-like receptor 11; Toll-like receptor 12 | |
Synthetic peptide taken within amino acid region 430-460 on human TLR11 protein. | |
200 μL | |
Primary | |
-20°C | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Chimpanzee, Human, Non-human Primate | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title