Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR12 Antibody (1229C), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB8086N
Description
TLR12 Monoclonal antibody specifically detects TLR12 in Mouse samples. It is validated for Western Blot, Flow CytometrySpecifications
TLR12 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
100131451 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Flow Cytometry | |
1229C | |
Western Blot, Flow Cytometry | |
toll-like receptor 12 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse TLR12, Thr22-Lys709, Accession # Q6QNU9 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction