Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR2 Mouse, Clone: TLR2/221, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2477470.1MG
Description
TLR2 Monoclonal specifically detects TLR2 in Human, Canine, Monkey samples. It is validated for Functional.Specifications
TLR2 | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready | |
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
Mouse | |
90 kDa | |
0.1 mg | |
Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
7097 | |
Human, Canine, Monkey | |
Purified |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
TLR2/221 | |
Unconjugated | |
PBS with No Preservative | |
TLR2 | |
Recombinant human TLR2 protein | |
Protein A or G purified | |
RUO | |
Primary | |
This MAb reacts with human Toll-like receptor 2 (TLR2). It is a member of the Toll-like receptor (TLR) family, which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is expressed most abundantly in peripheral blood leukocytes, and mediates host response to Gram-positive bacteria and yeast via stimulation of NF-kappaB. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction