Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR2 Rabbit anti-Human, DyLight 680, Clone: 67, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289250FR
Description
TLR2 Monoclonal specifically detects TLR2 in Human samples. It is validated for Flow Cytometry.Specifications
TLR2 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
67 | |
Flow Cytometry | |
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human TLR2 (NP_003255.2; Met1-Arg587). | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
7097 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title