Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR2 Rat anti-Mouse, Alexa Fluor™ 532, Clone: 11G5, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP227166AF532

Catalog No. NP227166A32


Add to cart

Description

Description

TLR2 Monoclonal antibody specifically detects TLR2 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

TLR2
11G5
Western Blot, Flow Cytometry
Conjugated, Purified
TLR2
A portion of amino acids 400-450 of murine TLR2 was used as the immunogen for this antibody.
Protein G purified
Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary
7097.0
Flow Cytometry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
Rat
IgG2a Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit