Learn More
TLR3 Rabbit anti-Human, Alexa Fluor™ 594, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP227167AF594
Catalog No. NP227167A94
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
TLR3 Polyclonal antibody specifically detects TLR3 in Human samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
TLR3
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
TLR3
The immunogen for this antibody was a portion of amino acids 325-393 of mouse TLR3.
0.1 mL
Primary
7098
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
CD283, CD283 antigen, toll-like receptor 3
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
Human