Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR3 Mouse anti-Mouse, Clone: PaT3, MilliporeSigma™

Mouse Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen TLR3
Clone PaT3
Concentration 1.0 mg/mL
Applications Flow Cytometry, Immunoprecipitation
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF2205100
View Documents
MilliporeSigma
MABF2205100UG
100 μg
Each for $373.00
Add to cart
 
MABF220525
View Documents
MilliporeSigma
MABF220525UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

TLR3 Monoclonal antibody specifically detects TLR3 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation.
Specifications

Specifications

TLR3
1.0 mg/mL
Unconjugated
Mouse
Mouse
Ba/F3 cells expressing Flag-6×His-conjugated mouse TLR3. Tlr3-/- mice were immunized.
Monoclonal
Protein G purified
PaT3
Flow Cytometry, Immunoprecipitation
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1κ
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit