Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA112823
Description
A suggested positive control for this product is placenta or pancreas lysate.
Toll-like receptors (TLRs) are evolutionarily conserved patternrecognition molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. The signaling of these TLRs is kept under tight control by the expression of endogenous inhibiting proteins such as RP105, a recently identified homolog of TLR4. This protein, in association with MD-1, interacts with and inhibits the TLR4/MD-2 signaling pathway.. While MD-2 can directly bind to LPS, one of the activating molecules of TLR4, the function of MD -1 is less well-known. It has been suggested however, that the RP105/MD-1 complex influences antibody production mediated by both TLR4/MD-2 and TLR2 receptor complexes.
Specifications
TLR3 | |
Polyclonal | |
PBS with 0.02% sodium azide | |
O15455, Q99MB1 | |
TLR3 | |
Synthetic peptide corresponding to 15 residues near the N-terminus of human TLR3. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
TLR3 | |
Toll-like Receptor 3, IIAE2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
142980, 7098 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction