Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR3 Rabbit anti-Human, Alexa Fluor™ 532, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227167AF532

Catalog No. NP227167A32


Add to cart

Description

Description

TLR3 Polyclonal antibody specifically detects TLR3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

TLR3
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CD283, CD283 antigen, toll-like receptor 3
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
TLR3
The immunogen for this antibody was a portion of amino acids 325-393 of mouse TLR3.
Protein G purified
Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Necroptosis, Signal Transduction, Toll Like Receptors
Primary
7098.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit