Learn More
TLR3 Rat anti-Human, Mouse, Alexa Fluor™ 350, Clone: 27N3D4, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP227404AF350
Catalog No. NP227404A30
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
TLR3 Monoclonal antibody specifically detects TLR3 in Human,Mouse samples. It is validated for Western Blot,Flow Cytometry.
Specifications
Specifications
TLR3
27N3D4
Western Blot, Flow Cytometry
CD283, CD283 antigen, toll-like receptor 3
Rat
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Flow Cytometry, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
TLR3
A portion of amino acids 350-375 of mouse TLR3 was used as the immunogen for this antibody.
0.1 mL
Primary
7098