Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR3 Rat anti-Human, Mouse, Alexa Fluor™ 532, Clone: 27N3D4, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227404AF532

Catalog No. NP227404A32


Add to cart

Description

Description

TLR3 Monoclonal antibody specifically detects TLR3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

TLR3
27N3D4
Western Blot, Flow Cytometry
Conjugated, Purified
TLR3
A portion of amino acids 350-375 of mouse TLR3 was used as the immunogen for this antibody.
Protein G purified
Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Necroptosis, Signal Transduction, Toll Like Receptors
Primary
7098.0
Flow Cytometry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CD283, CD283 antigen, toll-like receptor 3
Rat
IgG2b Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit