Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR4 Mouse anti-Human, Mouse, Rat, Porcine, Bovine, Mammalia, Alexa Fluor 532, Clone: 76B357.1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227149AF532
Description
TLR4 Monoclonal specifically detects TLR4 in Human, Mouse, Rat, Porcine, Bovine, Mammal samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Knockout Validated, Knockdown Validated.Specifications
TLR4 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
TLR4 | |
This antibody was developed against a portion of amino acids 100-200 of human TLR4 (NP_612564). | |
0.1 mL | |
Primary | |
The amino acid sequence used has 100% identity in human, chimp, baboon, Chinese hamster, and rat; 93% identity in bovine and pig; 85% identity in sheep and 78% identity in cat, horse and mouse. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, KnockDown | |
76B357.1 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Knockout Validated, KnockDown Validated | |
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
Mouse | |
Protein G purified | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
7099.0 | |
Human, Mouse, Rat, Porcine, Bovine, Mammalia | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title