Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR4 Rabbit anti-Human, FITC, Polyclonal, FabGennix

Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc TLR4FITC

Description
TLR4 Polyclonal antibody specifically detects TLR4 in Human samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
TLR4 | |
Polyclonal | |
FITC | |
TLR4 | |
ARMD10; CD284; EGK_07381; homolog of Drosophila toll; hToll; lipopolysaccharide response; Lps; Ly87; Ran/M1; Rasl2-8; Tlr4; TLR-4; TOLL; toll like receptor 4; toll/interleukin-l receptor (TIR) domain; toll4; toll-like receptor 4; toll-like receptor 4 immunity-related protein; Toll-like receptor 4-like protein; Toll-like receptor4 protein | |
Synthetic peptide taken within amino acid region 970-1000 on human TLR4 protein. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
O00206 | |
Rabbit | |
Affinity chromatography | |
RUO | |
7099 | |
-20° C, store in dark | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title