Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR4 Rabbit anti-Human, Rat, Porcine, Canine, Guinea Pig, Monkey, Clone: TLR4/3895R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen TLR4
Clone TLR4/3895R
Concentration 0.2 mg/ml
Dilution ELISA
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122931
View Documents
Novus Biologicals
NBP30772910UG
100 μg
Each for $406.52
Add to cart
 
NB122932
View Documents
Novus Biologicals
NBP30772920UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

TLR4 Monoclonal antibody specifically detects TLR4 in Human, Rat, Porcine, Canine, Guinea Pig, Monkey samples. It is validated for ELISA
Specifications

Specifications

TLR4
0.2 mg/ml
ELISA
Purified
RUO
Human, Rat, Porcine, Canine, Guinea Pig, Monkey
ARMD10, CD_antigen: CD284, CD284, CD284 antigen, EC 3.2.2.6, EC:3.2.2.6, homolog of Drosophila toll, hToll, TLR4, TLR-4, TOLL, toll like receptor 4 protein, toll-like receptor 4
Recombinant full-length human TLR4 protein.
Monoclonal
Protein A or G purified
TLR4/3895R
ELISA
Unconjugated
Rabbit
Adaptive Immunity, Autophagy, Cytokine Research, Immunology, Inflammation, Innate Immunity, Neurodegeneration, Signal Transduction, Virology Bacteria and Parasites
10 mM PBS with 0.05% BSA
7099
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit