Learn More
TLR4 Rat anti-Mouse, Alexa Fluor™ 350, Clone: MTS510, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP224865AF350
Catalog No. NP22486A30
Add to cart
Description
Description
TLR4 Monoclonal antibody specifically detects TLR4 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
TLR4
MTS510
Flow Cytometry, Flow (Cell Surface)
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4
Rat
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
In FA, the antibody blocks activation of monocytes with LPS (Akashi et al. 2000). This antibody preferentially recognizes TLR4-MD-2 complex than of TLR4 alone. The optimal condition has to be determined for individual experiments.
Flow Cytometry
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
TLR4
This antibody was developed by immunizing rats with the Ba/F3 cell line expressing mouse TLR4 and MD-2 (Akashi et al, 2000).
0.1 mL
Primary
7099
Mouse